accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrOHBr

CAS RN 13979-81-2

IUPAC Name: 3,5-dibromo-4-methylphenol
Synonyms: 3,5-dibromo-4-methylphenol
Molecular Weight (g/mol): 265.93
Molecular Formula: C7H6Br2O
InChi Key: AXCQKKVGMZCWPC-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(O)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results