accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13984-57-1

IUPAC Name: ethyl 5-oxohexanoate
Synonyms: ethyl 5-oxohexanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: MGPSIDGTLFKDEY-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCC(C)=O
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)