CAS RN 13986-26-0
IUPAC Name:
zirconium(4+) tetraiodide
Synonyms:
zirconium(IV) iodide
Molecular Weight (g/mol):
598.84
Molecular Formula:
I4Zr
InChi Key:
XLMQAUWIRARSJG-UHFFFAOYSA-J
SMILES:
[Zr+4].[I-].[I-].[I-].[I-]
1
–
1
of
1
results
1
Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals
CAS: 13986-26-0 Molecular Formula: I4Zr MDL Number: MFCD00049641 Synonym: zirconium iodide,unii-h6nzc1810v,zirconium iv ; iodide,acmc-1bzrf,zirconium 4+ tetraiodide