accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13986-53-3

IUPAC Name: λ²-cobalt(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Synonyms: λ²-cobalt(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular Weight (g/mol): 425.48
Molecular Formula: C22H38CoO4
InChi Key: KLJJOSZRALJWDS-UHFFFAOYSA-N
SMILES: [Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results