accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FBOHOH

CAS RN 139911-27-6

IUPAC Name: (4-fluoro-3-methylphenyl)boronic acid
Synonyms: 4-fluoro-3-methylphenylboronic acid
Molecular Weight (g/mol): 153.95
Molecular Formula: C7H8BFO2
InChi Key: JCIJCHSRVPSOML-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1F)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
  • (2)