accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FBHOOH

CAS RN 139911-29-8

IUPAC Name: (4-fluoro-2-methylphenyl)boronic acid
Synonyms: 4-fluoro-2-methylphenylboronic acid
Molecular Weight (g/mol): 153.95
Molecular Formula: C7H8BFO2
InChi Key: IQMLIVUHMSIOQP-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1B(O)O
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)