CAS RN 139926-23-1
IUPAC Name:
3-ethoxythiophene-2-carboxylate
Synonyms:
3-ethoxythiophene-2-carboxylate
Molecular Weight (g/mol):
171.19
Molecular Formula:
C7H7O3S
InChi Key:
FTKDCOINUAYGTC-UHFFFAOYSA-M
SMILES:
CCOC1=C(SC=C1)C([O-])=O
1
–
1
of
1
results
1
Promotions Available
3-Ethoxythiophene-2-carboxylic acid, 97%
3-Ethoxythiophene-2-carboxylic acid, 97%
CAS: 139926-23-1 Molecular Formula: C7H8O3S Molecular Weight (g/mol): 172.198 MDL Number: MFCD00205205 InChI Key: FTKDCOINUAYGTC-UHFFFAOYSA-N PubChem CID: 2777652 IUPAC Name: 3-ethoxythiophene-2-carboxylic acid SMILES: CCOC1=C(SC=C1)C(=O)O