accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(E)

CAS RN 140-10-3

IUPAC Name: (2E)-3-phenylprop-2-enoic acid
Synonyms: cinnamic acid 3t-phenyl-acrylic acid acidum cinnamylicum cinnamic acid, 14C-labeled cpd cinnamic acid, nickel (+2) salt trans-zimtsaeure 13C-labeled cpd cinnamic acid cinnamic acid, nickel(II) salt cinnamic acid, (z)-isomer nickel (+2) salt cinnamic acid
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: WBYWAXJHAXSJNI-VOTSOKGWSA-N
SMILES: OC(=O)\C=C\C1=CC=CC=C1
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (10)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (3)
  • (5)
  • (3)
Physical Form 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (5)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (5)
  • (10)
  • (1)
  • (1)
  • (4)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)