CAS RN 140-22-7
1,5-Diphenylcarbazide, ACS
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 MDL Number: MFCD00003013 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
sym-Diphenylcarbazide, 98%, ACS reagent
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.28 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
1,5-Diphenylcarbazide, 97+%
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 MDL Number: MFCD00003013 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
1,5-Diphenylcarbohydrazide, ACS, Spectrum™ Chemical
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.28 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N IUPAC Name: 1,3-bis(phenylamino)urea SMILES: O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
Diphenylcarbazide, 0.5% in Acetone, for Chromium, Certified, LabChem™
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
Diphenylcarbazide, ACS Grade, LabChem™
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
Diphenylcarbazide, 1% (w/v) Alcoholic, Ricca Chemical
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.28 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-bis(phenylamino)urea SMILES: O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
1,5-Diphenylcarbonohydrazide 98.0+%, TCI America™
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 MDL Number: MFCD00003013 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
1,5-Diphenylcarbohydrazide, ACS Reagent Grade, Ricca Chemical
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
Diphenylcarbazide, 0.5% (w/v) in Acetone, Ricca Chemical
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
Diphenylcarbazide, 1% Alcoholic, Certified, LabChem™
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.28 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-bis(phenylamino)urea SMILES: O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
Sigma Aldrich P,P-Dichlorophenylphosphine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1,5-Diphenylcarbazide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1,5-Diphenylcarbazide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
1,5-Diphenylcarbazide (Powder/Certified ACS), Fisher Chemical
CAS: 140-22-7 Molecular Formula: C13H14N4O Molecular Weight (g/mol): 242.282 MDL Number: MFCD00003013 InChI Key: KSPIHGBHKVISFI-UHFFFAOYSA-N Synonym: 1,5-diphenylcarbazide,diphenylcarbazide,1,5-diphenylcarbohydrazide,carbonic dihydrazide, 2,2'-diphenyl,1,5-diphenylcarbonohydrazide,1,5-diphenylcabohydrazide,sym-diphenylcarbazide,n,n'-diphenylcarbazide,2,2'-diphenylcarbazide,carbohydrazide, 1,5-diphenyl PubChem CID: 8789 ChEBI: CHEBI:4641 IUPAC Name: 1,3-dianilinourea SMILES: C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2