accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 140-67-0

IUPAC Name: 1-methoxy-4-(prop-2-en-1-yl)benzene
Synonyms: tarragon fema number 2411 allylanisole estragole epa pesticide chemical code 062150 chavicyl methyl ether 4-methoxyallylbenzene p-methoxyallylbenzene estragole (natural)
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: ZFMSMUAANRJZFM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC=C)C=C1
Percent Purity 
  • (1)
  • (2)
  • (5)
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)