accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 140-77-2

IUPAC Name: 3-cyclopentylpropanoic acid
Synonyms: cyclopentanepropionic acid cyclopentylproprionic acid
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: ZRPLANDPDWYOMZ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1CCCC1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (7)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)