accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CCH3NH2

CAS RN 140-80-7

IUPAC Name: (4-aminopentyl)diethylamine
Synonyms: (4-aminopentyl)diethylamine
Molecular Weight (g/mol): 158.29
Molecular Formula: C9H22N2
InChi Key: CAPCBAYULRXQAN-UHFFFAOYNA-N
SMILES: CCN(CC)CCCC(C)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results