CAS RN 140-82-9
IUPAC Name:
2-[2-(diethylamino)ethoxy]ethan-1-ol
Synonyms:
2-[2-(diethylamino)ethoxy]ethanol
Molecular Weight (g/mol):
161.25
Molecular Formula:
C8H19NO2
InChi Key:
VKBVRNHODPFVHK-UHFFFAOYSA-N
SMILES:
CCN(CC)CCOCCO
1
–
1
of
1
results
1
2-[2-(Diethylamino)ethoxy]ethanol 98.0+%, TCI America™
CAS: 140-82-9 Molecular Formula: C8H19NO2 Molecular Weight (g/mol): 161.25 MDL Number: MFCD00020604 InChI Key: VKBVRNHODPFVHK-UHFFFAOYSA-N Synonym: Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether PubChem CID: 67327 IUPAC Name: 2-[2-(diethylamino)ethoxy]ethan-1-ol SMILES: CCN(CC)CCOCCO