accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14003-16-8

IUPAC Name: 1-(5-methylfuran-2-yl)methanamine
Synonyms: 1-(5-methylfuran-2-yl)methanamine
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: YSEAGSCGERFGBL-UHFFFAOYSA-N
SMILES: CC1=CC=C(CN)O1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)