accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14014-88-1

IUPAC Name: ruthenium(3+) tribromide
Synonyms: ruthenium(3+) tribromide
Molecular Weight (g/mol): 340.78
Molecular Formula: Br3Ru
InChi Key: WYRXRHOISWEUST-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Ru+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results