accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14017-52-8

H2OGdOClOOOOClOOOOClOOO

CAS RN 14017-52-8

IUPAC Name: gadolinium(3+) hydrate triperchlorate
Synonyms: gadolinium(3+) hydrate triperchlorate
Molecular Weight (g/mol): 473.6
Molecular Formula: Cl3GdH2O13
InChi Key: TXABYLVQTHIELE-UHFFFAOYSA-K
SMILES: O.[Gd+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)