accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14017-54-0

IUPAC Name: holmium(3+) hexahydrate triperchlorate
Synonyms: holmium(3+) hexahydrate triperchlorate
Molecular Weight (g/mol): 571.36
Molecular Formula: Cl3H12HoO18
InChi Key: RRHAFXDVTMGQFK-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Ho+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results