accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14024-18-1

IUPAC Name: iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Synonyms: iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 353.17
Molecular Formula: C15H21FeO6
InChi Key: AQBLLJNPHDIAPN-LNTINUHCSA-K
SMILES: [Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (7)
  • (3)
  • (6)
  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (6)