accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14024-63-6

IUPAC Name: zinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: zinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 263.6
Molecular Formula: C10H14O4Zn
InChi Key: CYDXJXDAFPJUQE-FDGPNNRMSA-L
SMILES: [Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)