accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOOOOOOOZnNaNa

CAS RN 14025-21-9

IUPAC Name: zinc(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: zinc(2+) disodium ion(4-) edta
Molecular Weight (g/mol): 399.57
Molecular Formula: C10H12N2Na2O8Zn
InChi Key: UBLOJEHIINPTTG-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)