accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONNHN

CAS RN 14028-44-5

IUPAC Name: 13-chloro-10-(piperazin-1-yl)-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene
Synonyms: amoxapine amoxapine (substance) 阿莫沙平 amoxapine cyanamid brand asendis desmethylloxapine wyeth brand of amoxapine demolox lederle brand of amoxapine moxadil
Molecular Weight (g/mol): 313.79
Molecular Formula: C17H16ClN3O
InChi Key: QWGDMFLQWFTERH-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC3=CC=CC=C3N=C(N3CCNCC3)C2=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)