accessibility menu, dialog, popup

UserName

Viewing profile as user:
CuOOOCCCH3CH3H3CH3CCH3H3CH3CCH3H3CCH3CH3H3CHHO

CAS RN 14040-05-2

IUPAC Name: copper(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Synonyms: copper(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular Weight (g/mol): 430.09
Molecular Formula: C22H38CuO4
InChi Key: JGMGCTZGOAONPQ-UHFFFAOYSA-N
SMILES: [Cu++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)