accessibility menu, dialog, popup

UserName

Viewing profile as user:
WCOCOCOCOCOCO

CAS RN 14040-11-0

IUPAC Name: tungsten hexacarbonyl
Synonyms: tungsten hexacarbonyl
Molecular Weight (g/mol): 351.9
Molecular Formula: C6O6W
InChi Key: FQNHWXHRAUXLFU-UHFFFAOYSA-N
SMILES: [W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)