accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOONNNOOCH3OOCH3OOH3C

CAS RN 140428-81-5

IUPAC Name: [3,4,5-tris(acetyloxy)-6-(2-azidoethoxy)oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-(2-azidoethoxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 417.37
Molecular Formula: C16H23N3O10
InChi Key: DPAXDKFCBLGMIZ-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OCCN=[N+]=[N-])C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results