accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 140472-69-1

IUPAC Name: 3-bromo-5-hydroxybenzoic acid
Synonyms: 3-bromo-5-hydroxybenzoic acid
Molecular Weight (g/mol): 217.02
Molecular Formula: C7H5BrO3
InChi Key: WGIBEMRBLBGETQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=CC(Br)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)