accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)OO(R)HO(S)CH3(S)O(R)O(R)CH3(S)O(S)(R)H3COH(S)OH(S)CH3O(R)(R)HONH3CCH3(R)O(R)H3CO(R)O(R)O(R)CH3(R)OH(R)OCH3(R)OCH3CH3OHPOHOHO(E)(E)

CAS RN 1405-53-4

IUPAC Name: 2-[(4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-{[(2R,3R,4R,5S,6R)-5-{[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde; phosphoric acid
Synonyms: phosphoric acid; tylosin
Molecular Weight (g/mol): 1014.11
Molecular Formula: C46H80NO21P
InChi Key: NBOODGNJLRRJNA-IAGPQMRQSA-N
SMILES: OP(O)(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(\C)=C\[C@@H]1CO[C@@H]1O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]1OC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results