CAS RN 1405-89-6
IUPAC Name:
4-(2-{[2-(1-amino-2-methylbutyl)-4,5-dihydro-1,3-thiazol-4-yl]formamido}-4-methylpentanamido)-4-[(1-{[18-(3-aminopropyl)-12-benzyl-15-(butan-2-yl)-3-(carbamoylmethyl)-6-(carboxymethyl)-9-[(4H-imidazol-4-yl)methyl]-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclopentacosan-21-yl]carbamoyl}-2-methylbutyl)carbamoyl]butanoic acid zinc
Synonyms:
bacitracin
Molecular Weight (g/mol):
1488.09
Molecular Formula:
C66H103N17O16SZn
InChi Key:
QSNOBVJFKSQBBD-UHFFFAOYNA-N
SMILES:
[Zn].CCC(C)C(N)C1=NC(CS1)C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(C(C)CC)C(=O)NC1CCCCNC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(CC2C=NC=N2)NC(=O)C(CC2=CC=CC=C2)NC(=O)C(NC(=O)C(CCCN)NC1=O)C(C)CC
1
–
2
of
2
results
1
Bacitracin Zinc Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
2
Promotions Available
Thermo Scientific Chemicals Bacitracin, zinc salt, potency min. 60 Units/mg
Thermo Scientific Chemicals Bacitracin, zinc salt, potency min. 60 Units/mg
CAS: 1405-89-6 MDL Number: MFCD16661228 Synonym: Zinc bacitracin