accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14062-24-9

IUPAC Name: ethyl 2-(4-chlorophenyl)acetate
Synonyms: ethyl 2-(4-chlorophenyl)acetate
Molecular Weight (g/mol): 198.65
Molecular Formula: C10H11ClO2
InChi Key: UTWBWFXECVFDPZ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)