accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFNClFCl

CAS RN 140623-89-8

IUPAC Name: 2,6-dichloro-1-fluoropyridin-1-ium; tetrafluoroboranuide
Synonyms: 2,6-dichloro-1-fluoropyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 253.79
Molecular Formula: C5H3BCl2F5N
InChi Key: JVYBYXWHPIEUOT-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.F[N+]1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results