accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClOOCH3

CAS RN 14064-10-9

IUPAC Name: 1,3-diethyl 2-chloropropanedioate
Synonyms: 1,3-diethyl 2-chloropropanedioate
Molecular Weight (g/mol): 194.61
Molecular Formula: C7H11ClO4
InChi Key: WLWCQKMQYZFTDR-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Cl)C(=O)OCC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results