accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 140675-42-9

IUPAC Name: 1-(3,5-difluoro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3,5-difluoro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: MCDJUVXLLXTCFP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=CC(F)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results