CAS RN 140695-84-7
CAS RN 140695-84-7
IUPAC Name:
                                            
                                                    tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
                                            
                                        
                                    
                                    
                                    
                                    
                                        Synonyms:
                                        
                                                tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
                                        
                                    
                                                Molecular Weight (g/mol):
                                                
                                                           215.29
                                                
                                            
                                            
                                                 Molecular Formula:
                                                
                                                        C11H21NO3
                                            
                                                
                                            
                                        InChi Key:
                                                
                                                        OJCLHERKFHHUTB-UHFFFAOYNA-N
                                                
                                            
                                            
                                            SMILES:
                                                
                                                            CC(C)(C)OC(=O)N1CCCC(CO)C1
                                                
                                            
                                        
                                        1
                                        –
                                        1
                                        of
                                        1
                                        results
                                    
                                
                                        1 
                                    
                            
                                    
                                                        Thermo Scientific Chemicals (S)-1-BOC-3-(Hydroxymethyl)piperidine, 97%, Thermo Scientific™
                                                
                                                
                                                
                                                    
                                                
                                                    
                                                
                                                                        CAS: 140695-84-7 Molecular Formula: C11H21NO3 Molecular Weight (g/mol): 215.29 InChI Key: OJCLHERKFHHUTB-VIFPVBQESA-N Synonym: s-1-boc-3-hydroxymethyl piperidine,s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate,s-n-boc-3-piperidinemethanol,tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate,s-1-n-boc-3-hydroxymethyl-piperidine,s-1-boc-3-hyroxymethyl piperidine,s-1-n-boc-3-hydroxymethyl piperidine,s-n-boc-3-hyroxymethyl piperidine,s-1-boc-3-hydroxymethylpiperidine PubChem CID: 1514448 IUPAC Name: tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCCC(C1)CO
