accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFEu

CAS RN 14077-39-5

IUPAC Name: λ²-europium(2+) difluoride
Synonyms: λ²-europium(2+) difluoride
Molecular Weight (g/mol): 189.96
Molecular Formula: EuF2
InChi Key: YLDGIMHVWSTRML-UHFFFAOYSA-L
SMILES: [F-].[F-].[Eu++]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results