accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14091-08-8

IUPAC Name: 2-amino-3-(4-chlorophenyl)propanoic acid
Synonyms: fenclonine fenclonine hydrochloride fenclonine (L)- fenclonina P-chlorophenylalanine,DL 4-chloro-3-phenylalanine p-clorophenylalanine (D)-fenclonine fenclonine, (D)- fenchlonine
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: NIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(Cl)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)