accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3C(Z)

CAS RN 141-05-9

IUPAC Name: 1,4-diethyl (2Z)-but-2-enedioate
Synonyms: diethyl maleate
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: IEPRKVQEAMIZSS-WAYWQWQTSA-N
SMILES: CCOC(=O)\C=C/C(=O)OCC
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)