accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3COSiCH3H3COSiCH3H3COSiCH3CH3H3C

CAS RN 141-62-8

IUPAC Name: 2,2,4,4,6,6,8,8-octamethyl-3,5,7-trioxa-2,4,6,8-tetrasilanonane
Synonyms: decamethyltetrasiloxane
Molecular Weight (g/mol): 310.69
Molecular Formula: C10H30O3Si4
InChi Key: YFCGDEUVHLPRCZ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results