accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOCH3OCH3ONH3CH3C(E)

CAS RN 141-66-2

IUPAC Name: (2E)-3-[(dimethoxyphosphoryl)oxy]-N,N-dimethylbut-2-enamide
Synonyms: dicrotophos caswell no. 376 diapadrin carbicron epa pesticide chemical code 035201 karbicron ektafos bidrin dicrotofos shell SD-3562
Molecular Weight (g/mol): 237.19
Molecular Formula: C8H16NO5P
InChi Key: VEENJGZXVHKXNB-VOTSOKGWSA-N
SMILES: COP(=O)(OC)O\C(C)=C\C(=O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results