accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141-75-3

IUPAC Name: butanoyl chloride
Synonyms: butyryl chloride
Molecular Weight (g/mol): 106.55
Molecular Formula: C4H7ClO
InChi Key: DVECBJCOGJRVPX-UHFFFAOYSA-N
SMILES: CCCC(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)