CAS RN 141-85-5
CAS RN 141-85-5
IUPAC Name:
hydrogen 3-chloroaniline chloride
Synonyms:
hydrogen 3-chloroaniline chloride
Molecular Weight (g/mol):
164.03
Molecular Formula:
C6H7Cl2N
InChi Key:
NMGHWHCTRGZZOP-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC1=CC=CC(Cl)=C1
1
–
1
of
1
results
1
3-Chloroaniline Hydrochloride 99.0+%, TCI America™
CAS: 141-85-5 Molecular Formula: C6H7Cl2N Molecular Weight (g/mol): 164.029 MDL Number: MFCD00035485 InChI Key: NMGHWHCTRGZZOP-UHFFFAOYSA-N Synonym: 3-chloroaniline hydrochloride,3-chloroanilinium chloride,devol orange c,devol orange gc,m-chloroaniline hydrochloride,orange gc salt,orange base ngc,orange gcs salt,orange salt ngc,ansibases orange gc PubChem CID: 67336 IUPAC Name: 3-chloroaniline;hydrochloride SMILES: C1=CC(=CC(=C1)Cl)N.Cl