accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141-91-3

IUPAC Name: 2,6-dimethylmorpholine
Synonyms: morpholine, 2,6-dimethyl-
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: HNVIQLPOGUDBSU-UHFFFAOYNA-N
SMILES: CC1CNCC(C)O1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)