accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14113-01-0

IUPAC Name: 8-ethoxy-8-oxooctanoic acid
Synonyms: 8-ethoxy-8-oxooctanoic acid
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: KCFVQEAPUOVXTH-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results