accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141179-72-8

IUPAC Name: 4-fluoro-2-(trifluoromethyl)benzoate
Synonyms: 4-fluoro-2-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 207.1
Molecular Formula: C8H3F4O2
InChi Key: JUHPDXOIGLHXTC-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(F)C=C1C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)