accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14125-70-3

IUPAC Name: (2R,4aR,8R,8aS)-2-phenyl-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-ol
Synonyms: (2R,4aR,8R,8aS)-2-phenyl-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-ol
Molecular Weight (g/mol): 234.25
Molecular Formula: C13H14O4
InChi Key: XMDUTBYCCVWPLD-FVCCEPFGSA-N
SMILES: O[C@@H]1C=CO[C@@H]2CO[C@H](O[C@@H]12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results