accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14126-37-5

IUPAC Name: dibromonickel; bis(triphenylphosphane)
Synonyms: nickel bromide; bis(triphenylphosphine)
Molecular Weight (g/mol): 743.09
Molecular Formula: C36H30Br2NiP2
InChi Key: QEKXARSPUFVXIX-UHFFFAOYSA-L
SMILES: Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results