CAS RN 1412905-42-0
IUPAC Name:
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms:
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol):
265.09
Molecular Formula:
C13H17BFNO3
InChi Key:
RBEAFMIZDXOHAT-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O
1
–
1
of
1
results
1
Promotions Available
5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™