accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14130-05-3

IUPAC Name: nonadecan-1-amine
Synonyms: nonadecan-1-amine
Molecular Weight (g/mol): 283.54
Molecular Formula: C19H41N
InChi Key: INAMEDPXUAWNKL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results