accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOHFF

CAS RN 141428-47-9

IUPAC Name: 2-(2,3-difluoro-6-nitrophenyl)acetic acid
Synonyms: (2,3-difluoro-6-nitrophenyl)acetic acid
Molecular Weight (g/mol): 217.13
Molecular Formula: C8H5F2NO4
InChi Key: KZNVCGPCWKYPKD-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(C=CC(F)=C1F)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)