accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOOOH3CCl

CAS RN 141433-60-5

IUPAC Name: 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-13-ylium chloride
Synonyms: berberine chloride
Molecular Weight (g/mol): 371.82
Molecular Formula: C20H18ClNO4
InChi Key: VKJGBAJNNALVAV-UHFFFAOYSA-M
SMILES: [Cl-].COC1=C(OC)C2=C[N+]3=C(C=C2C=C1)C1=CC2=C(OCO2)C=C1CC3
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)