accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141474-37-5

IUPAC Name: 2,4-dibromo-6-fluoroaniline
Synonyms: 2,4-dibromo-6-fluoroaniline
Molecular Weight (g/mol): 268.91
Molecular Formula: C6H4Br2FN
InChi Key: YJLXEKFYZIBUPJ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results