accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1415-73-2

IUPAC Name: (10R)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracen-9-one
Synonyms: barbaloin barbaloin, β-d-isomer barbaloin, monoglucoside (S)-isomer barbaloin isobarbaloin barbaloin, β-D- barbaloin, β-d- barbaloin, (S)-isomer aloinum aloin A
Molecular Weight (g/mol): 418.4
Molecular Formula: C21H22O9
InChi Key: AFHJQYHRLPMKHU-WEZNYRQKSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)